ENA018109254 Enamine

5-cyclopropyl-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03986056
  • Cas Number : 733790-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06064

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06064
Availability details for
Quantity
  • Availability
  • Add to Cart