ENA018109256 Enamine

4-(chloromethyl)-6,7-dihydroxy-2H-chromen-2-one

  • MDL number MFCD03987203
  • Cas Number : 85029-91-0
  • Molecular Weight : 226.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06067
Availability details for
Quantity
  • Availability
  • Add to Cart