ENA018109267 Enamine

N-{[4-(methylsulfanyl)phenyl]methylidene}hydroxylamine

  • MDL number MFCD01043211
  • Cas Number : 1817768-41-4
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06084
Availability details for
Quantity
  • Availability
  • Add to Cart