ENA018109287 Enamine

(3,5-Dimethyl-isoxazol-4-ylmethyl)-phenyl-amine

  • MDL number MFCD04124300
  • Cas Number : 733796-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06120

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06120
Availability details for
Quantity
  • Availability
  • Add to Cart