ENA018109289 Enamine

3-({2-[(2-methoxyphenyl)carbamoyl]phenyl}carbamoyl)propanoic acid

  • MDL number MFCD03988315
  • Cas Number : 571156-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-128
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06123
Availability details for
Quantity
  • Availability
  • Add to Cart