ENA018109290 Enamine

2-amino-N-(4-methylphenyl)benzamide

  • MDL number MFCD00115975
  • Cas Number : 32212-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06124
Availability details for
Quantity
  • Availability
  • Add to Cart