ENA018109292 Enamine

3-ethoxy-4-[2-(morpholin-4-yl)-2-oxoethoxy]benzaldehyde

  • MDL number MFCD03987221
  • Cas Number : 98840-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06128
Availability details for
Quantity
  • Availability
  • Add to Cart