ENA018109300 Enamine

4-(3,5-Dimethyl-phenyl)-1-mercapto-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-one

  • MDL number MFCD03987289
  • Cas Number : 565178-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06139
Availability details for
Quantity
  • Availability
  • Add to Cart