ENA018109303 Enamine

1-(2,4-Dimethyl-phenyl)-1H-imidazole-2-thiol

  • MDL number MFCD03987294
  • Cas Number : 26209-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06142

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06142
Availability details for
Quantity
  • Availability
  • Add to Cart