ENA018109344 Enamine

7-methoxy-4-phenyl-1,2-dihydroquinolin-2-one

  • MDL number MFCD00572408
  • Cas Number : 30034-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06186
Availability details for
Quantity
  • Availability
  • Add to Cart