ENA018109369 Enamine

ethyl 5-carbamoyl-2-(2-chloroacetamido)-4-methylthiophene-3-carboxylate

  • MDL number MFCD01354258
  • Cas Number : 551910-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06220
Availability details for
Quantity
  • Availability
  • Add to Cart