ENA018109379 Enamine

1-[2-(5-Furan-2-yl-4-phenyl-4H-[1,2,4]triazol-3-ylsulfanyl)-acetyl]-piperidine-4-carboxylic acid

  • MDL number MFCD03988933
  • Cas Number : 735336-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06231

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06231
Availability details for
Quantity
  • Availability
  • Add to Cart