ENA018109387 Enamine

N-(3-methylphenyl)-4-(4-nitrophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD01239127
  • Cas Number : 300852-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06239
Availability details for
Quantity
  • Availability
  • Add to Cart