ENA018109391 Enamine

1-Butyl-5-(morpholine-4-sulfonyl)-1H-benzoimidazole-2-thiol

  • MDL number MFCD03988233
  • Cas Number : 735320-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06243

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06243
Availability details for
Quantity
  • Availability
  • Add to Cart