ENA018109398 Enamine

3-(2-sulfanyl-1H-1,3-benzodiazol-1-yl)propanenitrile

  • MDL number MFCD02679766
  • Cas Number : 56172-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-157
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06254
Availability details for
Quantity
  • Availability
  • Add to Cart