ENA018109410 Enamine

2-Chloro-5-(pyrrolidine-1-sulfonyl)-pyridine

  • MDL number MFCD03988229
  • Cas Number : 64614-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06266
Availability details for
Quantity
  • Availability
  • Add to Cart