ENA018109427 Enamine

2-(4-bromo-2-fluorophenoxy)acetic acid

  • MDL number MFCD03989628
  • Cas Number : 451-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-120
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06283
Availability details for
Quantity
  • Availability
  • Add to Cart