ENA018109434 Enamine

3-[(benzylsulfanyl)methyl]-1-benzofuran-2-carboxylic acid

  • MDL number MFCD03986073
  • Cas Number : 565174-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06290
Availability details for
Quantity
  • Availability
  • Add to Cart