ENA018109435 Enamine

3-[(cyclohexylsulfanyl)methyl]-1-benzofuran-2-carboxylic acid

  • MDL number MFCD03986074
  • Cas Number : 568559-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06291
Availability details for
Quantity
  • Availability
  • Add to Cart