ENA018109436 Enamine

3-{[(4-chlorophenyl)sulfanyl]methyl}-1-benzofuran-2-carboxylic acid

  • MDL number MFCD03986075
  • Cas Number : 733790-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06292
Availability details for
Quantity
  • Availability
  • Add to Cart