ENA018109483 Enamine

4-Morpholin-4-yl-benzaldehyde oxime

  • MDL number MFCD03217066
  • Cas Number : 433322-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06345

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06345
Availability details for
Quantity
  • Availability
  • Add to Cart