ENA018109486 Enamine

3-(2-chloroacetyl)-1-(2-methylpropyl)urea

  • MDL number MFCD04606929
  • Cas Number : 560995-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06348
Availability details for
Quantity
  • Availability
  • Add to Cart