ENA018109488 Enamine

5-Chloromethyl-furan-2-carboxylic acid isopropyl ester

  • MDL number MFCD04621462
  • Cas Number : 90416-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06350

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06350
Availability details for
Quantity
  • Availability
  • Add to Cart