ENA018109493 Enamine

5-(2-hydroxybenzoyl)-2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD04621556
  • Cas Number : 730950-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06355
Availability details for
Quantity
  • Availability
  • Add to Cart