ENA018109497 Enamine

3-(4-nitrophenyl)-1,2-oxazol-5-amine

  • MDL number MFCD04621564
  • Cas Number : 119162-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06361
Availability details for
Quantity
  • Availability
  • Add to Cart