ENA018109498 Enamine

ethyl 2-(2-chloroacetamido)-4-(2,4-dimethylphenyl)thiophene-3-carboxylate

  • MDL number MFCD02854001
  • Cas Number : 730949-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06362

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06362
Availability details for
Quantity
  • Availability
  • Add to Cart