ENA018109502 Enamine

2-(1H-indol-3-yl)-2-phenylethan-1-amine

  • MDL number MFCD01567108
  • Cas Number : 5027-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-82
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06366
Availability details for
Quantity
  • Availability
  • Add to Cart