ENA018109504 Enamine

2-(4-ethoxyphenoxy)aniline

  • MDL number MFCD04606945
  • Cas Number : 565198-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06368
Availability details for
Quantity
  • Availability
  • Add to Cart