ENA018109505 Enamine

2-chloro-1-{2,5-dimethyl-1-[3-(trifluoromethyl)phenyl]-1H-pyrrol-3-yl}ethan-1-one

  • MDL number MFCD01917466
  • Cas Number : 314245-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-97
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06369
Availability details for
Quantity
  • Availability
  • Add to Cart