ENA018109515 Enamine

3-(3-methylbutyl)-4-oxo-3,4-dihydrophthalazine-1-carboxylic acid

  • MDL number MFCD03986481
  • Cas Number : 565174-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06379
Availability details for
Quantity
  • Availability
  • Add to Cart