ENA018109517 Enamine

4-amino-5-[(3,4-dimethoxyphenyl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04606931
  • Cas Number : 568570-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-164
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06381
Availability details for
Quantity
  • Availability
  • Add to Cart