ENA018109529 Enamine

5-[(4-ethylphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD04606914
  • Cas Number : 565198-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-152
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06393
Availability details for
Quantity
  • Availability
  • Add to Cart