ENA018109540 Enamine

7-(Piperidine-1-sulfonyl)-3,4-dihydro-1H-quinoxalin-2-one

  • MDL number MFCD04606966
  • Cas Number : 556007-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06407

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06407
Availability details for
Quantity
  • Availability
  • Add to Cart