ENA018109563 Enamine

4-(4-bromophenyl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-1,3-thiazol-2-amine

  • MDL number MFCD03148703
  • Cas Number : 364740-29-4
  • Molecular Weight : 419.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06432
Availability details for
Quantity
  • Availability
  • Add to Cart