ENA018109569 Enamine

N-(6-Amino-1-butyl-2,4-dioxo-1,2,3,4-tetrahydro-pyrimidin-5-yl)-N-butyl-4-chloro-butyramide

  • MDL number MFCD04621522
  • Cas Number : 730950-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06438

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06438
Availability details for
Quantity
  • Availability
  • Add to Cart