ENA018109601 Enamine

2-(chloromethyl)-N,N-diethyl-1H-1,3-benzodiazole-6-sulfonamide

  • MDL number MFCD04621530
  • Cas Number : 730950-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06477
Availability details for
Quantity
  • Availability
  • Add to Cart