ENA018109602 Enamine

1-butyl-2-methyl-5-(morpholine-4-sulfonyl)-1H-1,3-benzodiazole

  • MDL number MFCD04608507
  • Cas Number : 736188-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06478
Availability details for
Quantity
  • Availability
  • Add to Cart