ENA018109625 Enamine

2-chloro-N,N'-bis(4-ethoxyphenyl)ethanimidamide

  • Cas Number : 40403-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06504
Availability details for
Quantity
  • Availability
  • Add to Cart