ENA018109631 Enamine

N-(2-methylphenyl)-2-(3-oxo-1,2,3,4-tetrahydroquinoxalin-2-yl)acetamide

  • MDL number MFCD01163327
  • Cas Number : 317814-58-7
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-06510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06510
Availability details for
Quantity
  • Availability
  • Add to Cart