ENA018109634 Enamine

2-(difluoromethoxy)-3-methoxybenzaldehyde

  • MDL number MFCD04621474
  • Cas Number : 730949-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-31
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06514
Availability details for
Quantity
  • Availability
  • Add to Cart