ENA018109643 Enamine

ethyl 6-amino-1-oxo-2H,3H,4H-1lambda4-thieno[2,3-b]thiopyran-5-carboxylate

  • MDL number MFCD04621536
  • Cas Number : 956452-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06523
Availability details for
Quantity
  • Availability
  • Add to Cart