ENA018109653 Enamine

N-[4-(chloromethyl)-2-oxo-2H-chromen-7-yl]acetamide

  • MDL number MFCD04610166
  • Cas Number : 447398-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-195
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06533
Availability details for
Quantity
  • Availability
  • Add to Cart