ENA018109655 Enamine

3-Amino-5-methyl-5-phenyl-imidazolidine-2,4-dione

  • MDL number MFCD04610171
  • Cas Number : 71202-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06535

Safety Information

msds https://www.enaminestore.com/catalog/EN300-06535
Availability details for
Quantity
  • Availability
  • Add to Cart