ENA018109662 Enamine

5-(4-Bromo-phenyl)-4H-[1,2,4]triazole-3-thiol

  • MDL number MFCD00708370
  • Cas Number : 39631-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225...227
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06542

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-06542
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart