ENA018109668 Enamine

4-(2,4-dichlorophenyl)-N-propyl-1,3-thiazol-2-amine

  • MDL number MFCD04608665
  • Cas Number : 556009-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06548
Availability details for
Quantity
  • Availability
  • Add to Cart