ENA018109670 Enamine

5-(propan-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04608652
  • Cas Number : 59689-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-186
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-06550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06550
Availability details for
Quantity
  • Availability
  • Add to Cart