ENA018109685 Enamine

1-({4-chloro-5,6-dimethylthieno[2,3-d]pyrimidin-2-yl}methyl)pyrrolidine

  • MDL number MFCD04610238
  • Cas Number : 733794-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-66
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06566
Availability details for
Quantity
  • Availability
  • Add to Cart