ENA018109687 Enamine

3-[cyclohexyl(methyl)sulfamoyl]benzoic acid

  • MDL number MFCD04605907
  • Cas Number : 554426-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06568
Availability details for
Quantity
  • Availability
  • Add to Cart