ENA018109689 Enamine

3-(propylsulfamoyl)benzoic acid

  • MDL number MFCD04607184
  • Cas Number : 7326-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-174
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06570
Availability details for
Quantity
  • Availability
  • Add to Cart