ENA018109701 Enamine

3-amino-5-ethyl-5-methylimidazolidine-2,4-dione

  • MDL number MFCD04612076
  • Cas Number : 1005-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06586
Availability details for
Quantity
  • Availability
  • Add to Cart